missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 90+%, ACS reagent

Catalog No. p-57099
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC400850250 25 g Glass bottle
AC400851000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC400850250 Supplier Thermo Scientific Chemicals Supplier No. 40085 0250
Only null left

CAS: 116-63-2 | C10H9NO4S | 239.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 116-63-2
Molecular Formula C10H9NO4S
Molecular Weight (g/mol) 239.25
MDL Number MFCD00004019
InChI Key RXCMFQDTWCCLBL-UHFFFAOYSA-N
Synonym 1-amino-2-naphthol-4-sulfonic acid, 4-amino-3-hydroxy-1-naphthalenesulfonic acid, 1-amino-4-sulfo-2-naphthol, 1,2,4-acid, unii-w46sk3p33g, 1-naphthalenesulfonic acid, 4-amino-3-hydroxy, 4-amino-3-hydroxynaphthalene-1-sulphonic acid, 4-amino-3-hydroxynaphthalenesulfonic acid, 1,2,4-aminonaphtholsulfonic acid, 2-hydroxy-4-sulfo-1-naphthylamine
PubChem CID 8316
ChEBI CHEBI:19024
IUPAC Name 4-amino-3-hydroxynaphthalene-1-sulfonic acid
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O

Specifications

Melting Point 295°C
Color Purple
Ignition Residue ≤0.1%
Infrared Spectrum Conforms to Ref Report
Assay Percent Range 90+%
Packaging Glass bottle
Linear Formula H2NC10H5(OH)SO3H
Quantity 25 g
Beilstein 14, 846
Sensitivity Pass Test (To Phosphates)
Merck Index 15, 450
Solubility Information (in sodium carbonate) Passes test
Formula Weight 239.25
Percent Purity 90+%
Grade ACS Reagent
Physical Form Powder
Chemical Name or Material 4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-147-3
RTECSNumber QK1292000
TSCA TSCA

RUO – Research Use Only