missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydroxy-1-naphthalene Sulfonic Acid, ACS, 90%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor A111625GM

 View more versions of this product

Catalog No. 18608258

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

116-63-2
0.001
C10H9NO4S
RXCMFQDTWCCLBL-UHFFFAOYSA-N
4-amino-3-hydroxynaphthalene-1-sulfonic acid
0.9
1
Poly Bottle
25 g
NC1=C2C=CC=CC2=C(C=C1O)S(O)(=O)=O
239.25
ACS

Chemical Identifiers

116-63-2
239.25
4-amino-3-hydroxynaphthalene-1-sulfonic acid
C10H9NO4S
RXCMFQDTWCCLBL-UHFFFAOYSA-N
NC1=C2C=CC=CC2=C(C=C1O)S(O)(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.