missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydroxy-1-naphthalene Sulfonic Acid, ACS, 90%, Spectrum™ Chemical

Catalog No. p-7164971
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608257 5 g Amber Glass Bottle
18608258 25 g Poly Bottle
18608259 100 g Poly Bottle
18608260 1 kg Poly Bottle
18608261 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608257 Supplier Spectrum Chemical Mfg Cor Supplier No. A11165GM
Only null left

Chemical Identifiers

CAS 116-63-2
Molecular Formula C10H9NO4S
Molecular Weight (g/mol) 239.25
InChI Key RXCMFQDTWCCLBL-UHFFFAOYSA-N
IUPAC Name 4-amino-3-hydroxynaphthalene-1-sulfonic acid
SMILES NC1=C2C=CC=CC2=C(C=C1O)S(O)(=O)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 0.9
Grade ACS

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.