missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole 95.0+%, TCI America™

$141.67 - $460.43

Chemical Identifiers

CAS 1750-12-5
Molecular Formula C2H6N6S
Molecular Weight (g/mol) 146.172
MDL Number MFCD00003098
InChI Key RDIMQHBOTMWMJA-UHFFFAOYSA-N
Synonym 4-amino-3-hydrazino-5-mercapto-1,2,4-triazole, purpald, 4-amino-3-hydrazino-1,2,4-triazol-5-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, hydrazone, purpald r, 4-amino-5-hydrazino-1,2,4-triazole-3-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, 3-hydrazone, 4-amino-5-hydrazinyl-4h-1,2,4-triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 4-amino-5-hydrazinyl-2,4-dihydro, 4-amino-5-hydrazinyl-1,2,4-triazole-3-thiol
PubChem CID 2723946
IUPAC Name 4-amino-3-hydrazinyl-1H-1,2,4-triazole-5-thione
SMILES C1(=S)NN=C(N1N)NN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A09565G
View Documents
TCI America
A09565G
5 g N/A
Only null left
 
A095625G
View Documents
TCI America
A095625G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1750-12-5
146.172
RDIMQHBOTMWMJA-UHFFFAOYSA-N
2723946
C1(=S)NN=C(N1N)NN
C2H6N6S
MFCD00003098
4-amino-3-hydrazino-5-mercapto-1,2,4-triazole, purpald, 4-amino-3-hydrazino-1,2,4-triazol-5-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, hydrazone, purpald r, 4-amino-5-hydrazino-1,2,4-triazole-3-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, 3-hydrazone, 4-amino-5-hydrazinyl-4h-1,2,4-triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 4-amino-5-hydrazinyl-2,4-dihydro, 4-amino-5-hydrazinyl-1,2,4-triazole-3-thiol
4-amino-3-hydrazinyl-1H-1,2,4-triazole-5-thione

Specifications

1750-12-5
White
MFCD00003098
1325
RDIMQHBOTMWMJA-UHFFFAOYSA-N
4-amino-3-hydrazinyl-1H-1,2,4-triazole-5-thione
2723946
≥95.0% (T)
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole [for Determination of Aldehydes]
240°C
C2H6N6S
5 g
4-amino-3-hydrazino-5-mercapto-1,2,4-triazole, purpald, 4-amino-3-hydrazino-1,2,4-triazol-5-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, hydrazone, purpald r, 4-amino-5-hydrazino-1,2,4-triazole-3-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, 3-hydrazone, 4-amino-5-hydrazinyl-4h-1,2,4-triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 4-amino-5-hydrazinyl-2,4-dihydro, 4-amino-5-hydrazinyl-1,2,4-triazole-3-thiol
C1(=S)NN=C(N1N)NN
146.172
146.17
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1091

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'