Learn More
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole, 99+%, Thermo Scientific Chemicals
$95.68 - $372.48
Chemical Identifiers
| CAS | 1750-12-5 |
|---|---|
| Molecular Formula | C2H6N6S |
| Molecular Weight (g/mol) | 146.172 |
| MDL Number | MFCD00003098 |
| InChI Key | RDIMQHBOTMWMJA-UHFFFAOYSA-N |
| Synonym | 4-amino-3-hydrazino-5-mercapto-1,2,4-triazole, purpald, 4-amino-3-hydrazino-1,2,4-triazol-5-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, hydrazone, purpald r, 4-amino-5-hydrazino-1,2,4-triazole-3-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, 3-hydrazone, 4-amino-5-hydrazinyl-4h-1,2,4-triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 4-amino-5-hydrazinyl-2,4-dihydro, 4-amino-5-hydrazinyl-1,2,4-triazole-3-thiol |
| PubChem CID | 2723946 |
| IUPAC Name | 4-amino-3-hydrazinyl-1H-1,2,4-triazole-5-thione |
| SMILES | C1(=S)NN=C(N1N)NN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0098206
|
Thermo Scientific Chemicals
L0098206 |
5 g |
Each for $95.68
|
|
|||||
|
AAL0098214
|
Thermo Scientific Chemicals
L0098214 |
25 g |
Each for $372.48
|
|
|||||
Description
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole is a reagent used in the determination of aldehydes and other reactive chemicals.was used in purpald assay for determination of bacterial polysaccharides containing substituted or unsubstituted glycol in residues such as glycerol, ribitol, arabinitol, furanosyl galactose and sialic acid. It was used as chromogen during the determination of catalase activity in small tissue samples. It was used as reagent in determination of formaldehyde. It was used for derivatization of neutral monosaccharides during HPLC.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole is a reagent used in the determination of aldehydes and other reactive chemicals.was used in purpald assay for determination of bacterial polysaccharides containing substituted or unsubstituted glycol in residues such as glycerol, ribitol, arabinitol, furanosyl galactose and sialic acid. It was used as chromogen during the determination of catalase activity in small tissue samples. It was used as reagent in determination of formaldehyde. It was used for derivatization of neutral monosaccharides during HPLC.
Solubility
Soluble in Dimethyl sulfoxide. (DMSO)
Notes
Store at -20°C. Avoid contact with heat. Incompatible with oxidizing agents.
Chemical Identifiers
| 1750-12-5 | |
| 146.172 | |
| RDIMQHBOTMWMJA-UHFFFAOYSA-N | |
| 2723946 | |
| C1(=S)NN=C(N1N)NN |
| C2H6N6S | |
| MFCD00003098 | |
| 4-amino-3-hydrazino-5-mercapto-1,2,4-triazole, purpald, 4-amino-3-hydrazino-1,2,4-triazol-5-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, hydrazone, purpald r, 4-amino-5-hydrazino-1,2,4-triazole-3-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, 3-hydrazone, 4-amino-5-hydrazinyl-4h-1,2,4-triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 4-amino-5-hydrazinyl-2,4-dihydro, 4-amino-5-hydrazinyl-1,2,4-triazole-3-thiol | |
| 4-amino-3-hydrazinyl-1H-1,2,4-triazole-5-thione |
Specifications
| 1750-12-5 | |
| C2H6N6S | |
| 5 g | |
| 134867 | |
| Soluble in Dimethyl sulfoxide. (DMSO) | |
| C1(=S)NN=C(N1N)NN | |
| 146.172 | |
| 146.17 | |
| 4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole |
| ∽220°C (decomposition) | |
| MFCD00003098 | |
| UN1325 | |
| 4-amino-3-hydrazino-5-mercapto-1,2,4-triazole, purpald, 4-amino-3-hydrazino-1,2,4-triazol-5-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, hydrazone, purpald r, 4-amino-5-hydrazino-1,2,4-triazole-3-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, 3-hydrazone, 4-amino-5-hydrazinyl-4h-1,2,4-triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 4-amino-5-hydrazinyl-2,4-dihydro, 4-amino-5-hydrazinyl-1,2,4-triazole-3-thiol | |
| RDIMQHBOTMWMJA-UHFFFAOYSA-N | |
| 4-amino-3-hydrazinyl-1H-1,2,4-triazole-5-thione | |
| 2723946 | |
| ≥99% |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 217-135-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only