missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole, 99+%, Thermo Scientific Chemicals

Catalog No. AAL0098214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0098214 25 g
AAL0098206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0098214 Supplier Thermo Scientific Chemicals Supplier No. L0098214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole is a reagent used in the determination of aldehydes and other reactive chemicals.was used in purpald assay for determination of bacterial polysaccharides containing substituted or unsubstituted glycol in residues such as glycerol, ribitol, arabinitol, furanosyl galactose and sialic acid. It was used as chromogen during the determination of catalase activity in small tissue samples. It was used as reagent in determination of formaldehyde. It was used for derivatization of neutral monosaccharides during HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole is a reagent used in the determination of aldehydes and other reactive chemicals.was used in purpald assay for determination of bacterial polysaccharides containing substituted or unsubstituted glycol in residues such as glycerol, ribitol, arabinitol, furanosyl galactose and sialic acid. It was used as chromogen during the determination of catalase activity in small tissue samples. It was used as reagent in determination of formaldehyde. It was used for derivatization of neutral monosaccharides during HPLC.

Solubility
Soluble in Dimethyl sulfoxide. (DMSO)

Notes
Store at -20°C. Avoid contact with heat. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1750-12-5
Molecular Formula C2H6N6S
Molecular Weight (g/mol) 146.172
MDL Number MFCD00003098
InChI Key RDIMQHBOTMWMJA-UHFFFAOYSA-N
Synonym 4-amino-3-hydrazino-5-mercapto-1,2,4-triazole, purpald, 4-amino-3-hydrazino-1,2,4-triazol-5-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, hydrazone, purpald r, 4-amino-5-hydrazino-1,2,4-triazole-3-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, 3-hydrazone, 4-amino-5-hydrazinyl-4h-1,2,4-triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 4-amino-5-hydrazinyl-2,4-dihydro, 4-amino-5-hydrazinyl-1,2,4-triazole-3-thiol
PubChem CID 2723946
IUPAC Name 4-amino-3-hydrazinyl-1H-1,2,4-triazole-5-thione
SMILES C1(=S)NN=C(N1N)NN

Specifications

Melting Point ∽220°C (decomposition)
Quantity 25 g
UN Number UN1325
Beilstein 134867
Solubility Information Soluble in Dimethyl sulfoxide. (DMSO)
Formula Weight 146.17
Percent Purity ≥99%
Chemical Name or Material 4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 217-135-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only