missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-fluorophenylboronic acid pinacol ester, 97%

Catalog No. AC444500010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444500010 1 g
AC444500050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC444500010 Supplier Thermo Scientific Chemicals Supplier No. 444500010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 819058-34-9 | C12H17BFNO2 | 237.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 819058-34-9
Molecular Formula C12H17BFNO2
Molecular Weight (g/mol) 237.08
MDL Number MFCD09033884
InChI Key AIXGNRNTXUKZLC-UHFFFAOYSA-N
Synonym 4-amino-3-fluorophenylboronic acid pinacol ester, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorophenylboronic acid, pinacol ester, benzenamine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-amino-3-fluorophenylboronicacidpinacolester, 4-amino-3-fluorophenylboronic acid,pinacol ester, 4-amino-3-fluorobenzeneboronic acid pinacol ester, 4-amino-3-fluorophenyl boronic acid pinacol ester, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-3-fluoro-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 17906172
IUPAC Name 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=CC(F)=C(N)C=C1

Specifications

Melting Point 101°C to 102°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 237.08
Percent Purity 97%
Chemical Name or Material 4-Amino-3-fluorophenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only