missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-3-fluorophenol 98.0+%, TCI America™
$132.35 - $549.89
Chemical Identifiers
| CAS | 399-95-1 |
|---|---|
| Molecular Formula | C6H6FNO |
| Molecular Weight (g/mol) | 127.118 |
| MDL Number | MFCD00077452 |
| InChI Key | MNPLTKHJEAFOCA-UHFFFAOYSA-N |
| Synonym | 3-fluoro-4-aminophenol, 2-fluoro-4-hydroxyaniline, phenol, 4-amino-3-fluoro, 4-amino-3-fluoro-phenol, pubchem2832, 3-fluoro-4-amino-phenol, 4-amino-3-fluorophenol, acmc-209j9q, 4-amine-3-fluorophenol, buttpark 33\04-79 |
| PubChem CID | 2735919 |
| IUPAC Name | 4-amino-3-fluorophenol |
| SMILES | C1=CC(=C(C=C1O)F)N |
Chemical Identifiers
| 399-95-1 | |
| 127.118 | |
| MNPLTKHJEAFOCA-UHFFFAOYSA-N | |
| 2735919 | |
| C1=CC(=C(C=C1O)F)N |
| C6H6FNO | |
| MFCD00077452 | |
| 3-fluoro-4-aminophenol, 2-fluoro-4-hydroxyaniline, phenol, 4-amino-3-fluoro, 4-amino-3-fluoro-phenol, pubchem2832, 3-fluoro-4-amino-phenol, 4-amino-3-fluorophenol, acmc-209j9q, 4-amine-3-fluorophenol, buttpark 33\04-79 | |
| 4-amino-3-fluorophenol |
Specifications
| 399-95-1 | |
| C6H6FNO | |
| 1 g | |
| 3-fluoro-4-aminophenol, 2-fluoro-4-hydroxyaniline, phenol, 4-amino-3-fluoro, 4-amino-3-fluoro-phenol, pubchem2832, 3-fluoro-4-amino-phenol, 4-amino-3-fluorophenol, acmc-209j9q, 4-amine-3-fluorophenol, buttpark 33\04-79 | |
| C1=CC(=C(C=C1O)F)N | |
| 127.118 | |
| 127.12 | |
| Crystalline Powder |
| 139°C | |
| MFCD00077452 | |
| 3077 | |
| MNPLTKHJEAFOCA-UHFFFAOYSA-N | |
| 4-amino-3-fluorophenol | |
| 2735919 | |
| ≥98.0% (GC,T) | |
| 4-Amino-3-fluorophenol |
Safety and Handling
TSCA : No