Learn More
4-Amino-3-fluorophenol, 98%, Thermo Scientific™
$178.47 - $448.73
Chemical Identifiers
| CAS | 399-95-1 |
|---|---|
| Molecular Formula | C6H6FNO |
| Molecular Weight (g/mol) | 127.12 |
| InChI Key | MNPLTKHJEAFOCA-UHFFFAOYSA-N |
| Synonym | 3-fluoro-4-aminophenol, 2-fluoro-4-hydroxyaniline, phenol, 4-amino-3-fluoro, 4-amino-3-fluoro-phenol, pubchem2832, 3-fluoro-4-amino-phenol, 4-amino-3-fluorophenol, acmc-209j9q, 4-amine-3-fluorophenol, buttpark 33\04-79 |
| PubChem CID | 2735919 |
| IUPAC Name | 4-amino-3-fluorophenol |
| SMILES | C1=CC(=C(C=C1O)F)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC458310010
|
N/A | 1g |
Each for $178.47
|
|
|||||
|
AC458310050
|
N/A | 5g |
Each for $448.73
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 399-95-1 | |
| 127.12 | |
| 3-fluoro-4-aminophenol, 2-fluoro-4-hydroxyaniline, phenol, 4-amino-3-fluoro, 4-amino-3-fluoro-phenol, pubchem2832, 3-fluoro-4-amino-phenol, 4-amino-3-fluorophenol, acmc-209j9q, 4-amine-3-fluorophenol, buttpark 33\04-79 | |
| 4-amino-3-fluorophenol |
| C6H6FNO | |
| MNPLTKHJEAFOCA-UHFFFAOYSA-N | |
| 2735919 | |
| C1=CC(=C(C=C1O)F)N |
Specifications
| 399-95-1 | |
| 97.5% min. (GC) | |
| C6H6FNO | |
| 3-fluoro-4-aminophenol, 2-fluoro-4-hydroxyaniline, phenol, 4-amino-3-fluoro, 4-amino-3-fluoro-phenol, pubchem2832, 3-fluoro-4-amino-phenol, 4-amino-3-fluorophenol, acmc-209j9q, 4-amine-3-fluorophenol, buttpark 33\04-79 | |
| MNPLTKHJEAFOCA-UHFFFAOYSA-N | |
| 4-amino-3-fluorophenol | |
| 2735919 | |
| 98% |
| Authentic | |
| Glass Bottle | |
| NH2C6H3(F)OH | |
| Solubility in water: slightly soluble. | |
| C1=CC(=C(C=C1O)F)N | |
| 127.12 | |
| 127.12 | |
| 4-Amino-3-fluorophenol |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
May cause cancer.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid relea
GHS Signal Word: Danger
RUO – Research Use Only