missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-fluorophenol, 98%

Catalog No. AC458310010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458310010 1 g
AC458310050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458310010 Supplier Thermo Scientific Chemicals Supplier No. 458310010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 399-95-1 | C6H6FNO | 127.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 399-95-1
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.12
InChI Key MNPLTKHJEAFOCA-UHFFFAOYSA-N
Synonym 3-fluoro-4-aminophenol, 2-fluoro-4-hydroxyaniline, phenol, 4-amino-3-fluoro, 4-amino-3-fluoro-phenol, pubchem2832, 3-fluoro-4-amino-phenol, 4-amino-3-fluorophenol, acmc-209j9q, 4-amine-3-fluorophenol, buttpark 33\04-79
PubChem CID 2735919
IUPAC Name 4-amino-3-fluorophenol
SMILES C1=CC(=C(C=C1O)F)N

Specifications

Melting Point 137.0°C to 139.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula NH2C6H3(F)OH
Quantity 1 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 127.12
Percent Purity 98%
Chemical Name or Material 4-Amino-3-fluorophenol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
May cause cancer.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid relea

RUO – Research Use Only