missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-ethylbenzonitrile, 96%

Catalog No. AAL1073203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1073203 1 g
AAL1073206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1073203 Supplier Thermo Scientific Chemicals Supplier No. L1073203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 170230-87-2 | C9H10N2 | 146.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 170230-87-2
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.193
MDL Number MFCD00041431
InChI Key YOHLABDNVQLZIA-UHFFFAOYSA-N
Synonym 4-amino-3-ethyl-benzonitrile, 4-cyano-2-ethylaniline, 4-amino-3-ethyl benzonitrile, buttpark 13\03-08, benzonitrile, 4-amino-3-ethyl-9ci, 4-amino-3-ethylbenzenecarbonitrile, pubchem16098, pubchem21039, acmc-209xcd, intermediates-zcf02269
PubChem CID 2735330
IUPAC Name 4-amino-3-ethylbenzonitrile
SMILES CCC1=C(C=CC(=C1)C#N)N

Specifications

Melting Point 60°C to 66°C
Quantity 1 g
UN Number UN3439
Beilstein 7526452
Formula Weight 146.19
Percent Purity 96%
Chemical Name or Material 4-Amino-3-ethylbenzonitrile
Hazard Category H302+H332-H311
Hazard Statement GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only