missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-chlorobenzotrifluoride 97.0+%, TCI America™

Catalog No. p-7117486
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15115G 5 g
A151125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15115G Supplier TCI America Supplier No. A15115G

Chemical Identifiers

CAS 39885-50-2
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.57
MDL Number MFCD00042563
InChI Key MBBUTABXEITVNY-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl aniline, 4-amino-3-chlorobenzotrifluoride, 4-amino-5-chloro-benzotrifluoride, benzenamine, 2-chloro-4-trifluoromethyl, 3-chloro-4-aminobenzotrifluoride, 2-chloro-4-trifluoromethyl-phenylamine, 2-chloro-4-trifluoromethyl benzenamine, 2-chloro-4-trifluoromethyl phenylamine, 4-amino-3-chloro-benzotrifluoride, pubchem2474
PubChem CID 162001
IUPAC Name 2-chloro-4-(trifluoromethyl)aniline
SMILES NC1=CC=C(C=C1Cl)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 195.57
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Amino-3-chlorobenzotrifluoride
EINECSNumber (3)-0266
TSCA No