missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-bromobenzonitrile 98.0+%, TCI America™

Catalog No. A19615G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19615G Supplier TCI America Supplier No. A19615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 50397-74-5
Melting Point 112°C
Color Yellow
Molecular Formula C7H5BrN2
Quantity 5 g
MDL Number MFCD01861386
UN Number 3439
Synonym 3-bromo-4-aminobenzonitrile, 2-bromo-4-cyanoaniline, benzonitrile, 4-amino-3-bromo, 4-amino-3-bromo-benzonitrile, 4-amino-3-brombenzonitrile, buttpark 35\03-64, 4-amino-3-bromobenzenecarbonitrile, pubchem4609, acmc-209klz, 4-amino-3-bromobenzonitile
InChI Key POESQIHWIIWNJL-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C#N)Br)N
IUPAC Name 4-amino-3-bromobenzonitrile
Molecular Weight (g/mol) 197.035
PubChem CID 1515279
Formula Weight 197.04
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-bromobenzonitrile
Show More Show Less