missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-bromobenzonitrile, 97%

Catalog No. p-7036979
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5548303 1 g
AAH5548306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5548303 Supplier Thermo Scientific Chemicals Supplier No. H5548303
Only null left

CAS: 50397-74-5 | C7H5BrN2 | 197.035 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50397-74-5
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD01861386
InChI Key POESQIHWIIWNJL-UHFFFAOYSA-N
Synonym 3-bromo-4-aminobenzonitrile, 2-bromo-4-cyanoaniline, benzonitrile, 4-amino-3-bromo, 4-amino-3-bromo-benzonitrile, 4-amino-3-brombenzonitrile, buttpark 35\03-64, 4-amino-3-bromobenzenecarbonitrile, pubchem4609, acmc-209klz, 4-amino-3-bromobenzonitile
PubChem CID 1515279
IUPAC Name 4-amino-3-bromobenzonitrile
SMILES C1=CC(=C(C=C1C#N)Br)N

Specifications

Melting Point 106°C to 110°C
Quantity 1 g
UN Number UN3439
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 4-Amino-3-bromobenzonitrile
Hazard Category H302+H312+H332-H315-H317-H319-H335-H500
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only