missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-bromo-2-chloropyridine, 97+%

Catalog No. AAH5235906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5235906 5 g
AAH5235903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5235906 Supplier Thermo Scientific Chemicals Supplier No. H5235906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 215364-85-5 | C5H4BrClN2 | 207.455 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 215364-85-5
Molecular Formula C5H4BrClN2
Molecular Weight (g/mol) 207.455
MDL Number MFCD09263478
InChI Key KTWAUKYSQFZIET-UHFFFAOYSA-N
Synonym 4-amino-3-bromo-2-chloropyridine, 3-bromo-2-chloro-4-pyridinamine, 3-bromo-2-chloro-4-pyridylamine, 3-bromo-2-chloro-pyridin-4-amine, 4-pyridinamine, 3-bromo-2-chloro, 4-amino-2-chloro-3-bromopyridine, pubchem2607, acmc-209flk, ksc547a7h, 3-bromo-2-chloro-4-aminopyridine
PubChem CID 29919300
IUPAC Name 3-bromo-2-chloropyridin-4-amine
SMILES C1=CN=C(C(=C1N)Br)Cl

Specifications

Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 207.46
Percent Purity ≥97%
Chemical Name or Material 4-Amino-3-bromo-2-chloropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only