CAS RN 215364-85-5
IUPAC Name:
3-bromo-2-chloropyridin-4-amine
Synonyms:
3-bromo-2-chloropyridin-4-amine
Molecular Weight (g/mol):
207.46
Molecular Formula:
C5H4BrClN2
InChi Key:
KTWAUKYSQFZIET-UHFFFAOYSA-N
SMILES:
NC1=C(Br)C(Cl)=NC=C1
1
–
1
of
1
results
1
Promotions Available
4-Amino-3-bromo-2-chloropyridine, 97+%
4-Amino-3-bromo-2-chloropyridine, 97+%
CAS: 215364-85-5 Molecular Formula: C5H4BrClN2 Molecular Weight (g/mol): 207.455 MDL Number: MFCD09263478 InChI Key: KTWAUKYSQFZIET-UHFFFAOYSA-N Synonym: 4-amino-3-bromo-2-chloropyridine,3-bromo-2-chloro-4-pyridinamine,3-bromo-2-chloro-4-pyridylamine,3-bromo-2-chloro-pyridin-4-amine,4-pyridinamine, 3-bromo-2-chloro,4-amino-2-chloro-3-bromopyridine,pubchem2607,acmc-209flk,ksc547a7h,3-bromo-2-chloro-4-aminopyridine PubChem CID: 29919300 IUPAC Name: 3-bromo-2-chloropyridin-4-amine SMILES: C1=CN=C(C(=C1N)Br)Cl