missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3,5-difluoropyridine, 97%

Catalog No. AC452530050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452530050 5 g
AC452530010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC452530050 Supplier Thermo Scientific Chemicals Supplier No. 452530050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 159783-22-9 | C5H4F2N2 | 130.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 159783-22-9
Molecular Formula C5H4F2N2
Molecular Weight (g/mol) 130.1
InChI Key XWAOYJHUAQFFMT-UHFFFAOYSA-N
PubChem CID 2737654
IUPAC Name 3,5-difluoropyridin-4-amine
SMILES C1=C(C(=C(C=N1)F)N)F

Specifications

Melting Point 102°C to 105°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Synonym 4-amino-3,5-difluoropyridine, 3,5-difluoro-4-pyridinamine, 3,5-difluoro-pyridin-4-ylamine, 4-pyridinamine, 3,5-difluoro, difluoropyridinamine, 3,5-difluoro-4-pyridylamine, 3,5-difluoropyridin-4-ylamine, 4-pyridinamine,3,5-difluoro-9ci, 4-pyridinamine, 3,5-difluoro-9ci
Formula Weight 130.1
Percent Purity 97%
Chemical Name or Material 4-Amino-3, 5-difluoropyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only