accessibility menu, dialog, popup

UserName

NFH2NF

CAS RN 159783-22-9

IUPAC Name: 3,5-difluoropyridin-4-amine
Synonyms: 3,5-difluoropyridin-4-amine
Molecular Weight (g/mol): 130.1
Molecular Formula: C5H4F2N2
InChi Key: XWAOYJHUAQFFMT-UHFFFAOYSA-N
SMILES: NC1=C(F)C=NC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results