missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3,5-dichlorobenzoic acid, 98%

Catalog No. AAA1498214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1498214 25 g
AAA1498206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1498214 Supplier Thermo Scientific Chemicals Supplier No. A1498214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56961-25-2 | C7H5Cl2NO2 | 206.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56961-25-2
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.02
MDL Number MFCD00017091
InChI Key UHXYYTSWBYTDPD-UHFFFAOYSA-N
Synonym 3,5-dichloro-4-aminobenzoic acid, unii-bik7ns3b2f, benzoic acid, 4-amino-3,5-dichloro, bik7ns3b2f, 4-amino-3,5-dichlorobenzoicacid, pubchem14451, acmc-209xmx, 4-14-00-01279 beilstein handbook reference, ksc495s0n, 4-carboxy-2,6-dichloroaniline
PubChem CID 42061
IUPAC Name 4-amino-3,5-dichlorobenzoic acid
SMILES NC1=C(Cl)C=C(C=C1Cl)C(O)=O

Specifications

Melting Point ∼290°C (decomposition)
Quantity 25 g
Beilstein 2805751
Formula Weight 206.03
Percent Purity 98%
Chemical Name or Material 4-Amino-3,5-dichlorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 260-468-9
RTECSNumber DG1930000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only