missing translation for 'onlineSavingsMsg'
Learn More

4'-Amino-3',5'-dichloroacetophenone 98.0+%, TCI America™

Catalog No. A19675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19675G 5 g
A196725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19675G Supplier TCI America Supplier No. A19675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37148-48-4
Molecular Formula C8H7Cl2NO
Molecular Weight (g/mol) 204.05
MDL Number MFCD00238535
InChI Key JLPKZJDZXIKSCP-UHFFFAOYSA-N
Synonym 4'-amino-3',5'-dichloroacetophenone, 4-amino-3,5-dichloroacetophenone, 1-4-amino-3,5-dichlorophenyl ethanone, 1-4-amino-3,5-dichlorophenyl ethan-1-one, 3,5-dichloro-4-aminoacetophenone, 1-4-amino-3,5-dichlorophenyl-ethanone, 4-acetyl-2,6-dichloroaniline, ethanone, 1-4-amino-3,5-dichlorophenyl, 1-acetyl-4-amino-3,5-dichlorobenzene, 1-4-amino-3,5-dichloro-phenyl-ethanone
PubChem CID 604812
IUPAC Name 1-(4-amino-3,5-dichlorophenyl)ethanone
SMILES CC(=O)C1=CC(=C(C(=C1)Cl)N)Cl

Specifications

Melting Point 165°C
Quantity 5 g
Formula Weight 204.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Amino-3′,5′-dichloroacetophenone