accessibility menu, dialog, popup

UserName

H3COClNH2Cl

CAS RN 37148-48-4

IUPAC Name: 1-(4-amino-3,5-dichlorophenyl)ethan-1-one
Synonyms: 1-(4-amino-3,5-dichlorophenyl)ethanone
Molecular Weight (g/mol): 204.05
Molecular Formula: C8H7Cl2NO
InChi Key: JLPKZJDZXIKSCP-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(Cl)=C(N)C(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results