missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-(trifluoromethyl)benzonitrile, 97%

Catalog No. AC442470050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442470050 5 g
AC442470010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC442470050 Supplier Thermo Scientific Chemicals Supplier No. 442470050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 654-70-6 | C8H5F3N2 | 186.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 654-70-6
Molecular Formula C8H5F3N2
Molecular Weight (g/mol) 186.14
InChI Key PMDYLCUKSLBUHO-UHFFFAOYSA-N
Synonym 4-amino-2-trifluoromethyl benzonitrile, 5-amino-2-cyanobenzotrifluoride, 4-cyano-3-trifluoromethylaniline, 2-cyano-5-aminobenzotrifluoride, 4-cyano-3-trifluoromethyl aniline, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08, 4-amino 2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile
PubChem CID 522170
IUPAC Name 4-amino-2-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1N)C(F)(F)F)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 144.0°C to 146.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 186.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Amino-2-(trifluoromethyl)benzonitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

RUO – Research Use Only