missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-(trifluoromethyl)benzonitrile, 97%

Catalog No. p-7045882
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1234603 1 g
AAL1234606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1234603 Supplier Thermo Scientific Chemicals Supplier No. L1234603

CAS: 654-70-6 | C8H5F3N2 | 186.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 654-70-6
Molecular Formula C8H5F3N2
Molecular Weight (g/mol) 186.137
MDL Number MFCD00042155
InChI Key PMDYLCUKSLBUHO-UHFFFAOYSA-N
Synonym 4-amino-2-trifluoromethyl benzonitrile, 5-amino-2-cyanobenzotrifluoride, 4-cyano-3-trifluoromethylaniline, 2-cyano-5-aminobenzotrifluoride, 4-cyano-3-trifluoromethyl aniline, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08, 4-amino 2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile
PubChem CID 522170
IUPAC Name 4-amino-2-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1N)C(F)(F)F)C#N

Specifications

Melting Point 136°C to 139°C
Quantity 1 g
UN Number UN3439
Beilstein 3278074
Formula Weight 186.14
Percent Purity 97%
Chemical Name or Material 4-Amino-2-(trifluoromethyl)benzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H331-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only