missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-(trifluoromethyl)benzoic acid, 97+%

Catalog No. p-7031519
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH3262603 1 g
AAH3262606 5 g
2 options

Catalog No. AAH3262603

Supplier: Thermo Scientific Chemicals H3262603

Only null left

CAS: 393-06-6 | C8H6F3NO2 | 205.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 393-06-6
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.14
MDL Number MFCD03407959
InChI Key AMVHEVZYTGHASE-UHFFFAOYSA-N
Synonym 4-amino-2-trifluoromethyl benzoic acid, 5-amino-2-carboxybenzotrifluoride, 2-trifluoromethyl-4-aminobenzoic acid, pubchem1360, 4-amimo-2-trifluoromethyl benzoicacid, acmc-209j4m, ksc495q5d, rarechem al bo 0438, buttpark 44\01-93, 4-amino-2-trifluoromethyl benzoicacid
PubChem CID 3836325
IUPAC Name 4-amino-2-(trifluoromethyl)benzoic acid
SMILES NC1=CC(=C(C=C1)C(O)=O)C(F)(F)F

Specifications

Melting Point 183°C to 186°C
Quantity 1 g
Formula Weight 205.14
Percent Purity ≥97%
Chemical Name or Material 4-Amino-2-(trifluoromethyl)benzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only