missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. A23495G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23495G 5 g
A234925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23495G Supplier TCI America Supplier No. A23495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2486-75-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD06656133
InChI Key XRSQZFJLEPBPOZ-UHFFFAOYSA-N
Synonym 4-amino-2-methyl-benzoic acid, 2-methyl-4-aminobenzoic acid, 2-methyl-4-amino-benzoic acid, 4-amino-2-methyl benzoic acid, benzoic acid, 4-amino-2-methyl, pubchem10924, 4-amino-o-toluic acid, methyl-p-aminobenzoesaure, acmc-1cnb3, methyl-4-amino benzoic acid
PubChem CID 241632
IUPAC Name 4-amino-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)N)C(=O)O

Specifications

Melting Point 164°C
Color Yellow
Quantity 5 g
Formula Weight 151.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-methylbenzoic Acid