missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-methyl-5-pyrimidinemethanol 98.0+%, TCI America™

Supplier:  TCI America A2730200MG

 View more versions of this product

Catalog No. A2730200MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Amino-2-methyl-5-pyrimidinemethanol
198°C
C6H9N3O
200 mg
VUTBELPREDJDDH-UHFFFAOYSA-N
(4-amino-2-methylpyrimidin-5-yl)methanol
777
139.16
Crystalline Powder
73-67-6
White-Yellow
MFCD00849239
4-Amino-5-hydroxymethyl-2-methylpyrimidine
CC1=NC=C(C(=N1)N)CO
139.158
CHEBI:16892
≥98.0% (GC,T)

Chemical Identifiers

73-67-6
139.158
VUTBELPREDJDDH-UHFFFAOYSA-N
777
(4-amino-2-methylpyrimidin-5-yl)methanol
C6H9N3O
MFCD00849239
4-Amino-5-hydroxymethyl-2-methylpyrimidine
CHEBI:16892
CC1=NC=C(C(=N1)N)CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions