missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-methyl-5-pyrimidinemethanol 98.0+%, TCI America™

Catalog No. p-7118046
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2730200MG 200 mg
A27301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2730200MG Supplier TCI America Supplier No. A2730200MG

Chemical Identifiers

CAS 73-67-6
Molecular Formula C6H9N3O
Molecular Weight (g/mol) 139.158
MDL Number MFCD00849239
InChI Key VUTBELPREDJDDH-UHFFFAOYSA-N
Synonym 4-Amino-5-hydroxymethyl-2-methylpyrimidine
PubChem CID 777
ChEBI CHEBI:16892
IUPAC Name (4-amino-2-methylpyrimidin-5-yl)methanol
SMILES CC1=NC=C(C(=N1)N)CO

Specifications

Melting Point 198°C
Color White-Yellow
Quantity 200 mg
Formula Weight 139.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-methyl-5-pyrimidinemethanol