missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-mercaptopyrimidine, 97%

Catalog No. p-2700536
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC214770010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC214770010 Supplier Thermo Scientific Chemicals Supplier No. 214770010
Only null left

CAS: 333-49-3 | C4H5N3S | 127.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 333-49-3
Molecular Formula C4H5N3S
Molecular Weight (g/mol) 127.17
InChI Key DCPSTSVLRXOYGS-UHFFFAOYSA-N
Synonym 2-thiocytosine, 4-amino-2-mercaptopyrimidine, thiocytosine, 4-aminopyrimidine-2-thiol, cytosine, thio, cytosine, 2-thio, 2 1h-pyrimidinethione, 4-amino, 2 1h-pyrimidinone, 4-aminothio, 4-amino-2 1h-pyrimidinethione, 4-amino-2-thiopyrimidine
PubChem CID 2724245
IUPAC Name 6-amino-1H-pyrimidine-2-thione
SMILES C1=C(NC(=S)N=C1)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 127.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Amino-2-mercaptopyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
EINECSNumber 206-374-3

RUO – Research Use Only