missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2-fluorobenzotrifluoride 98.0+%, TCI America™
$171.49 - $689.70
Chemical Identifiers
| CAS | 69411-68-3 |
|---|---|
| Molecular Formula | C7H5F4N |
| Molecular Weight (g/mol) | 179.12 |
| MDL Number | MFCD00190120 |
| InChI Key | CRRVZRDISHOQQL-UHFFFAOYSA-N |
| Synonym | 4-amino-2-fluorobenzotrifluoride, 3-fluoro-4-trifluoromethyl aniline, 2-fluoro-4-amino benzotrifluoride, benzenamine, 3-fluoro-4-trifluoromethyl, 3-fluoro-4-trifluoromethyl-phenylamine, 3-fluoro-4-trifluoromethyl phenylamine, pubchem1631, 4-amino,2-fluorobenzotrifluoride, 3-fluoro-4-trifluoromethyl-aniline |
| PubChem CID | 2735914 |
| IUPAC Name | 3-fluoro-4-(trifluoromethyl)aniline |
| SMILES | NC1=CC=C(C(F)=C1)C(F)(F)F |
Chemical Identifiers
| 69411-68-3 | |
| 179.12 | |
| CRRVZRDISHOQQL-UHFFFAOYSA-N | |
| 2735914 | |
| NC1=CC=C(C(F)=C1)C(F)(F)F |
| C7H5F4N | |
| MFCD00190120 | |
| 4-amino-2-fluorobenzotrifluoride, 3-fluoro-4-trifluoromethyl aniline, 2-fluoro-4-amino benzotrifluoride, benzenamine, 3-fluoro-4-trifluoromethyl, 3-fluoro-4-trifluoromethyl-phenylamine, 3-fluoro-4-trifluoromethyl phenylamine, pubchem1631, 4-amino,2-fluorobenzotrifluoride, 3-fluoro-4-trifluoromethyl-aniline | |
| 3-fluoro-4-(trifluoromethyl)aniline |
Specifications
| 69411-68-3 | |
| C7H5F4N | |
| 1 g | |
| CRRVZRDISHOQQL-UHFFFAOYSA-N | |
| 3-fluoro-4-(trifluoromethyl)aniline | |
| 2735914 | |
| ≥98.0% (GC,T) | |
| 4-Amino-2-fluorobenzotrifluoride |
| 56°C | |
| MFCD00190120 | |
| 4-amino-2-fluorobenzotrifluoride, 3-fluoro-4-trifluoromethyl aniline, 2-fluoro-4-amino benzotrifluoride, benzenamine, 3-fluoro-4-trifluoromethyl, 3-fluoro-4-trifluoromethyl-phenylamine, 3-fluoro-4-trifluoromethyl phenylamine, pubchem1631, 4-amino,2-fluorobenzotrifluoride, 3-fluoro-4-trifluoromethyl-aniline | |
| NC1=CC=C(C(F)=C1)C(F)(F)F | |
| 179.12 | |
| 179.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No