missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-fluorobenzotrifluoride 98.0+%, TCI America™

Catalog No. A24691G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24691G 1 g
A24695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24691G Supplier TCI America Supplier No. A24691G

Chemical Identifiers

CAS 69411-68-3
Molecular Formula C7H5F4N
Molecular Weight (g/mol) 179.12
MDL Number MFCD00190120
InChI Key CRRVZRDISHOQQL-UHFFFAOYSA-N
Synonym 4-amino-2-fluorobenzotrifluoride, 3-fluoro-4-trifluoromethyl aniline, 2-fluoro-4-amino benzotrifluoride, benzenamine, 3-fluoro-4-trifluoromethyl, 3-fluoro-4-trifluoromethyl-phenylamine, 3-fluoro-4-trifluoromethyl phenylamine, pubchem1631, 4-amino,2-fluorobenzotrifluoride, 3-fluoro-4-trifluoromethyl-aniline
PubChem CID 2735914
IUPAC Name 3-fluoro-4-(trifluoromethyl)aniline
SMILES NC1=CC=C(C(F)=C1)C(F)(F)F

Specifications

Melting Point 56°C
Quantity 1 g
Formula Weight 179.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-fluorobenzotrifluoride