missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-dimethylamino-6-hydroxypyrimidine Hemihydrate 98.0+%, TCI America™

Catalog No. A14691G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14691G Supplier TCI America Supplier No. A14691G

Chemical Identifiers

CAS 76750-84-0
Molecular Formula C6H10N4O
Molecular Weight (g/mol) 154.173
MDL Number MFCD00090845
InChI Key ZBWZIUPJSKQVQC-UHFFFAOYSA-N
Synonym 6-Amino-2-dimethylamino-4-pyrimidinol, 6-Amino-2-dimethylamino-4(1H)-pyrimidinone
PubChem CID 230048
IUPAC Name 6-amino-2-(dimethylamino)-1H-pyrimidin-4-one
SMILES CN(C)C1=NC(=O)C=C(N1)N

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 154.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-dimethylamino-6-hydroxypyrimidine Hemihydrate