accessibility menu, dialog, popup

UserName

H3CNCH3NOH2NHN

CAS RN 76750-84-0

IUPAC Name: 6-amino-2-(dimethylamino)-1,4-dihydropyrimidin-4-one
Synonyms: 6-amino-2-(dimethylamino)-1H-pyrimidin-4-one
Molecular Weight (g/mol): 154.17
Molecular Formula: C6H10N4O
InChi Key: ZBWZIUPJSKQVQC-UHFFFAOYSA-N
SMILES: CN(C)C1=NC(=O)C=C(N)N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results