missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chloro-6,7-dimethoxyquinazoline 98.0+%, TCI America™

$80.85 - $240.64

Chemical Identifiers

CAS 23680-84-4
Molecular Formula C10H10ClN3O2
Molecular Weight (g/mol) 239.66
MDL Number MFCD00051734
InChI Key HWIIAAVGRHKSOJ-UHFFFAOYSA-N
Synonym 4-amino-2-chloro-6,7-dimethoxyquinazoline, 2-chloro-4-amino-6,7-dimethoxyquinazoline, 2-chloro-6,7-dimethoxy-4-quinazolinamine, 2-chloro-4-amino-6,7-dimethoxy quinazoline, unii-fg7z4t71y4, 2-chloro-4-amino-6,7-dimethoxy-quinazoline, 4-quinazolinamine, 2-chloro-6,7-dimethoxy, 2-chloro-6,7-dimethoxy-4-aminoquinazoline, 2-chloro-6,7-dimethoxyquinazolin-4-ylamine, 4-amino-2-chloro-6,7-dimethoxy quinazoline
PubChem CID 90235
IUPAC Name 2-chloro-6,7-dimethoxyquinazolin-4-amine
SMILES COC1=C(OC)C=C2C(N)=NC(Cl)=NC2=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A21515G
View Documents
TCI America
A21515G
5 g
Each for $80.85
Only null left
 
A215125G
View Documents
TCI America
A215125G
25 g
Each for $240.64
Only null left
 
Description

Description

Specifications

Chemical Identifiers

23680-84-4
239.66
HWIIAAVGRHKSOJ-UHFFFAOYSA-N
90235
COC1=C(OC)C=C2C(N)=NC(Cl)=NC2=C1
C10H10ClN3O2
MFCD00051734
4-amino-2-chloro-6,7-dimethoxyquinazoline, 2-chloro-4-amino-6,7-dimethoxyquinazoline, 2-chloro-6,7-dimethoxy-4-quinazolinamine, 2-chloro-4-amino-6,7-dimethoxy quinazoline, unii-fg7z4t71y4, 2-chloro-4-amino-6,7-dimethoxy-quinazoline, 4-quinazolinamine, 2-chloro-6,7-dimethoxy, 2-chloro-6,7-dimethoxy-4-aminoquinazoline, 2-chloro-6,7-dimethoxyquinazolin-4-ylamine, 4-amino-2-chloro-6,7-dimethoxy quinazoline
2-chloro-6,7-dimethoxyquinazolin-4-amine

Specifications

23680-84-4
White-Yellow
MFCD00051734
4-amino-2-chloro-6,7-dimethoxyquinazoline, 2-chloro-4-amino-6,7-dimethoxyquinazoline, 2-chloro-6,7-dimethoxy-4-quinazolinamine, 2-chloro-4-amino-6,7-dimethoxy quinazoline, unii-fg7z4t71y4, 2-chloro-4-amino-6,7-dimethoxy-quinazoline, 4-quinazolinamine, 2-chloro-6,7-dimethoxy, 2-chloro-6,7-dimethoxy-4-aminoquinazoline, 2-chloro-6,7-dimethoxyquinazolin-4-ylamine, 4-amino-2-chloro-6,7-dimethoxy quinazoline
COC1=C(OC)C=C2C(N)=NC(Cl)=NC2=C1
239.66
239.66
Crystalline Powder
302°C
C10H10ClN3O2
5 g
HWIIAAVGRHKSOJ-UHFFFAOYSA-N
2-chloro-6,7-dimethoxyquinazolin-4-amine
90235
≥98.0% (HPLC,N)
4-Amino-2-chloro-6,7-dimethoxyquinazoline
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'