missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2-chloro-6,7-dimethoxyquinazoline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 302°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 239.66 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Amino-2-chloro-6,7-dimethoxyquinazoline |
Safety and Handling
| TSCA | No |