missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chloro-5-nitropyrimidine, 97%, Thermo Scientific Chemicals

Catalog No. p-7055805
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6447803 1 g
AAH6447806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6447803 Supplier Thermo Scientific Chemicals Supplier No. H6447803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1920-66-7 | Molecular Formula: C4H3ClN4O2 | Molecular Weight: 174.54 g/mol

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 1920-66-7
Molecular Formula C4H3ClN4O2
Molecular Weight (g/mol) 174.54
MDL Number MFCD00127771
InChI Key RZGOEIWDMVQJBQ-UHFFFAOYSA-N
PubChem CID 74716
IUPAC Name 2-chloro-5-nitropyrimidin-4-amine
SMILES NC1=NC(Cl)=NC=C1[N+]([O-])=O

Specifications

Melting Point 221°C to 226°C
Quantity 1 g
Beilstein 156953
Synonym 4-amino-2-chloro-5-nitropyrimidine, 4-pyrimidinamine, 2-chloro-5-nitro, 2-chloro-4-amino-5-nitropyrimidine, 2-chloro-5-nitro-pyrimidin-4-ylamine, 2-chloro-5-nitro-4-pyrimidinamine, 4-pyrimidinamine,2-chloro-5-nitro, acmc-209x0o, 4-amino-5-nitro-2-chloropyrimidine, 2-chloro-5-nitropyrimidine-4-amine
Solubility Information Slightly soluble in water.
Formula Weight 174.54
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Amino-2-chloro-5-nitropyrimidine
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 217-648-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only