accessibility menu, dialog, popup

UserName

NH2NClNNOO

CAS RN 1920-66-7

IUPAC Name: 2-chloro-5-nitropyrimidin-4-amine
Synonyms: 2-chloro-5-nitropyrimidin-4-amine
Molecular Weight (g/mol): 174.54
Molecular Formula: C4H3ClN4O2
InChi Key: RZGOEIWDMVQJBQ-UHFFFAOYSA-N
SMILES: NC1=NC(Cl)=NC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results