Learn More
4-Amino-2-chloro-5-nitropyrimidine, 97%, Thermo Scientific Chemicals
Description
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 221°C to 226°C |
| Quantity | 5 g |
| Beilstein | 156953 |
| Synonym | 4-amino-2-chloro-5-nitropyrimidine, 4-pyrimidinamine, 2-chloro-5-nitro, 2-chloro-4-amino-5-nitropyrimidine, 2-chloro-5-nitro-pyrimidin-4-ylamine, 2-chloro-5-nitro-4-pyrimidinamine, 4-pyrimidinamine,2-chloro-5-nitro, acmc-209x0o, 4-amino-5-nitro-2-chloropyrimidine, 2-chloro-5-nitropyrimidine-4-amine |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 174.54 |
| Percent Purity | 97% |
| Physical Form | Solid |
| Chemical Name or Material | 4-Amino-2-chloro-5-nitropyrimidine |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 217-648-7 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only