missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chloro-5-fluoropyrimidine 98.0+%, TCI America™

Catalog No. A28091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28091G 1 g
A28095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28091G Supplier TCI America Supplier No. A28091G

Chemical Identifiers

CAS 155-10-2
Molecular Formula C4H3ClFN3
Molecular Weight (g/mol) 147.537
MDL Number MFCD00057344
InChI Key SLQAJWTZUXJPNY-UHFFFAOYSA-N
Synonym 4-amino-2-chloro-5-fluoropyrimidine, 2-chloro-5-fluoro-pyrimidin-4-ylamine, 2-chloro-4-amino-5-fluoropyrimidine, 2-chloro-5-fluoro-4-aminopyrimidine, 2-chloro-5-fluoropyrimidin-4-ylamine, 2-chloro-5-fluoropyrimidine-4-ylamine, 4-pyrimidinamine, 2-chloro-5-fluoro, 4-pyrimidinamine, 2-chloro-5-fluoro-9ci, pubchem6930, 2-chlor-4-amino-5-fluorpyrimidin
PubChem CID 254372
IUPAC Name 2-chloro-5-fluoropyrimidin-4-amine
SMILES C1=C(C(=NC(=N1)Cl)N)F

Specifications

Melting Point 195°C
Color White
Quantity 1 g
Formula Weight 147.54
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-chloro-5-fluoropyrimidine