accessibility menu, dialog, popup

UserName

CAS RN 155-10-2

IUPAC Name: 2-chloro-5-fluoropyrimidin-4-amine
Synonyms: 2-chloro-5-fluoropyrimidin-4-amine
Molecular Weight (g/mol): 147.54
Molecular Formula: C4H3ClFN3
InChi Key: SLQAJWTZUXJPNY-UHFFFAOYSA-N
SMILES: NC1=NC(Cl)=NC=C1F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)