missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-butanol, 98%

Numéro de catalogue. p-3735268
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
Packaging:
Bouteille en verre
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC330230010 1 g Bouteille en verre
AC330230050 5 g Bouteille en verre
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC330230010 Fournisseur Thermo Scientific Chemicals Code fournisseur 330230010

CAS : 39884-48-5 | C4H11NO | 89,14 g/mol

Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.

Identifiants chimiques

CAS 39884-48-5
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00216936
InChI Key NAXUFNXWXFZVSI-UHFFFAOYSA-N
Synonym 4-amino-2-butanol, 2-butanol, 4-amino, 4-amino-butan-2-ol, 3-hydroxy-n-butylamine, 1-aminomethylpropan-2-ol, 4-aminobutan-2 r,s-ol
PubChem CID 170254
IUPAC Name 4-aminobutan-2-ol
SMILES CC(CCN)O

Spécifications

CAS Min % 97.5
CAS Max % 100.0
Color Incolore à jaune
Density 0.9300g/mL
Boiling Point 103.0°C to 104.0°C (34.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Bouteille en verre
Refractive Index 1.4518 to 1.4538
Linear Formula NH2(CH2)2CH(OH)CH3
Quantity 1 g
Specific Gravity 0.93
Formula Weight 89.14
Percent Purity 98%
Physical Form Liquide
Chemical Name or Material 4-Amino-2-butanol
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 254-673-2

RUO – Usage réservé à la recherche