accessibility menu, dialog, popup

UserName

CAS RN 39884-48-5

IUPAC Name: (3R)-3-hydroxybutan-1-aminium
Synonyms: (3R)-3-hydroxybutan-1-aminium
Molecular Weight (g/mol): 90.15
Molecular Formula: C4H12NO
InChi Key: NAXUFNXWXFZVSI-SCSAIBSYSA-O
SMILES: C[C@@H](O)CC[NH3+]
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)