missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-bromobenzonitrile, 95%

CAS: 53312-82-6 | C7H5BrN2 | 197.04 g/mol

$100.45 - $384.84

Chemical Identifiers

CAS 53312-82-6
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD03695814
InChI Key OSYGPHDNNWRFJO-UHFFFAOYSA-N
Synonym 2-bromo-4-aminobenzonitrile, ksc494a6n, benzonitrile, 4-amino-2-bromo
PubChem CID 824602
IUPAC Name 4-amino-2-bromobenzonitrile
SMILES NC1=CC(Br)=C(C=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6124703
View Documents
Thermo Scientific Chemicals
H6124703
1 g
Each for $100.45
Only null left
 
AAH6124706
View Documents
Thermo Scientific Chemicals
H6124706
5 g
Each for $384.84
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

53312-82-6
197.04
OSYGPHDNNWRFJO-UHFFFAOYSA-N
824602
NC1=CC(Br)=C(C=C1)C#N
C7H5BrN2
MFCD03695814
2-bromo-4-aminobenzonitrile, ksc494a6n, benzonitrile, 4-amino-2-bromo
4-amino-2-bromobenzonitrile

Specifications

53312-82-6
C7H5BrN2
1 g
2-bromo-4-aminobenzonitrile, ksc494a6n, benzonitrile, 4-amino-2-bromo
NC1=CC(Br)=C(C=C1)C#N
197.04
197.03
Powder
Yellow
MFCD03695814
UN3439
OSYGPHDNNWRFJO-UHFFFAOYSA-N
4-amino-2-bromobenzonitrile
824602
95%
4-Amino-2-bromobenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only