missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-bromobenzonitrile, 95%

Catalog No. AAH6124706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6124706 5 g
AAH6124703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6124706 Supplier Thermo Scientific Chemicals Supplier No. H6124706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 53312-82-6 | C7H5BrN2 | 197.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53312-82-6
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD03695814
InChI Key OSYGPHDNNWRFJO-UHFFFAOYSA-N
Synonym 2-bromo-4-aminobenzonitrile, ksc494a6n, benzonitrile, 4-amino-2-bromo
PubChem CID 824602
IUPAC Name 4-amino-2-bromobenzonitrile
SMILES NC1=CC(Br)=C(C=C1)C#N

Specifications

Color Yellow
Quantity 5 g
UN Number UN3439
Formula Weight 197.03
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Amino-2-bromobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only