Learn More
4-Amino-2,5-difluorobenzeneboronic acid pinacol ester, 96%
CAS: 939807-75-7 | C12H16BF2NO2 | 255.072 g/mol
Supplier: Thermo Scientific Chemicals H5235603
Description
4-Amino-2,5-difluorobenzeneboronic acid pinacol ester is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Amino-2,5-difluorobenzeneboronic acid pinacol ester is used as pharmaceutical intermediate.
Notes
Air sensitive. Store away from oxidizing agents.
Specifications
| 4-Amino-2,5-difluorobenzeneboronic acid pinacol ester | |
| C12H16BF2NO2 | |
| 1 g | |
| 4-amino-2,5-difluorobenzeneboronic acid pinacol ester, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylamine, 4-amino-2,5-difluorophenylboronic acid pinacol ester, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2F)N)F | |
| 255.072 | |
| 255.07 |
| 939807-75-7 | |
| MFCD09743045 | |
| Air Sensitive | |
| SOHBHFWRCLJGDV-UHFFFAOYSA-N | |
| 2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline | |
| 57955144 | |
| 96% |
Chemical Identifiers
| 939807-75-7 | |
| 255.072 | |
| SOHBHFWRCLJGDV-UHFFFAOYSA-N | |
| 57955144 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2F)N)F |
| C12H16BF2NO2 | |
| MFCD09743045 | |
| 4-amino-2,5-difluorobenzeneboronic acid pinacol ester, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylamine, 4-amino-2,5-difluorophenylboronic acid pinacol ester, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
| 2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only