missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,5-difluorobenzeneboronic acid pinacol ester, 96%

Catalog No. p-7034664
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52356MD 250 mg
AAH5235603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52356MD Supplier Thermo Scientific Chemicals Supplier No. H52356MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 939807-75-7 | C12H16BF2NO2 | 255.072 g/mol

4-Amino-2,5-difluorobenzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-2,5-difluorobenzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

Notes
Air sensitive. Store away from oxidizing agents.

Chemical Identifiers

CAS 939807-75-7
Molecular Formula C12H16BF2NO2
Molecular Weight (g/mol) 255.072
MDL Number MFCD09743045
InChI Key SOHBHFWRCLJGDV-UHFFFAOYSA-N
Synonym 4-amino-2,5-difluorobenzeneboronic acid pinacol ester, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylamine, 4-amino-2,5-difluorophenylboronic acid pinacol ester, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 57955144
IUPAC Name 2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2F)N)F

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 255.07
Percent Purity 96%
Chemical Name or Material 4-Amino-2,5-difluorobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only