missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,3,5,6-tetrafluorobenzoic Acid 97.0+%, TCI America™

Catalog No. A09871G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09871G 1 g
A09875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09871G Supplier TCI America Supplier No. A09871G

Chemical Identifiers

CAS 944-43-4
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD00007647
InChI Key WTNSXWSOTDBWOR-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluoro-4-aminobenzoic acid, acmc-209rrw, 4-amino-2,3,5,6-tetrafluorobenzoicacid, 2,3,5,6-tetrafluoro-4-amino-benzoic acid, 4-amino-2,3,5,6-tetrafluoro-benzoic acid, benzoic acid,4-amino-2,3,5,6-tetrafluoro, benzoic acid, 4-amino-2,3,5,6-tetrafluoro, 4-amino-2,3,5,6-tetrafluorobenzoic acid
PubChem CID 70345
IUPAC Name 4-amino-2,3,5,6-tetrafluorobenzoic acid
SMILES C1(=C(C(=C(C(=C1F)F)N)F)F)C(=O)O

Specifications

Melting Point 181°C
Quantity 1 g
Formula Weight 209.10
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2,3,5,6-tetrafluorobenzoic Acid