missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,3,5,6-tetrafluorobenzoic acid, 97%

Catalog No. AAL2012803
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2012803 1 g
AAL2012814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2012803 Supplier Thermo Scientific Chemicals Supplier No. L2012803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 944-43-4 | C7H3F4NO2 | 209.1 g/mol

It is used in chemical research and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in chemical research and as pharmaceutical intermediate.

Solubility
Insoluble in water. Soluble in methanol (25 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 944-43-4
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD00007647
InChI Key WTNSXWSOTDBWOR-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluoro-4-aminobenzoic acid, acmc-209rrw, 4-amino-2,3,5,6-tetrafluorobenzoicacid, 2,3,5,6-tetrafluoro-4-amino-benzoic acid, 4-amino-2,3,5,6-tetrafluoro-benzoic acid, benzoic acid,4-amino-2,3,5,6-tetrafluoro, benzoic acid, 4-amino-2,3,5,6-tetrafluoro, 4-amino-2,3,5,6-tetrafluorobenzoic acid
PubChem CID 70345
IUPAC Name 4-amino-2,3,5,6-tetrafluorobenzoic acid
SMILES C1(=C(C(=C(C(=C1F)F)N)F)F)C(=O)O

Specifications

Melting Point 182°C to 184°C
Quantity 1 g
Beilstein 2121117
Solubility Information Insoluble in water. Soluble in methanol (25mg/ml).
Formula Weight 209.11
Percent Purity 98%
Chemical Name or Material 4-Amino-2,3,5,6-tetrafluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-409-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only