missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-naphthalenesulfonic Acid 98.0+%, TCI America™

Catalog No. A034425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A034425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A034425G Supplier TCI America Supplier No. A034425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84-86-6
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.25
MDL Number MFCD00004027
InChI Key NRZRRZAVMCAKEP-UHFFFAOYSA-N
Synonym naphthionic acid, 4-amino-1-naphthalenesulfonic acid, piria's acid, 1-naphthylamine-4-sulfonic acid, naphthionsaeure, 1,4-naphthionic acid, sulfonaphtin, 1-amino-4-sulfonaphthalene, usaf m-5, 1-aminonaphthalene-4-sulfonic acid
PubChem CID 6790
ChEBI CHEBI:38219
IUPAC Name 4-aminonaphthalene-1-sulfonic acid
SMILES NC1=C2C=CC=CC2=C(C=C1)S(O)(=O)=O

Specifications

Color Red-White
Quantity 25 g
Formula Weight 223.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-1-naphthalenesulfonic Acid
EINECSNumber (4)-0491
RTECSNumber QK1270000
TSCA Yes