Learn More
4-Amino-1-butanol, 98%
CAS: 13325-10-5 | C4H11NO | 89.138 g/mol
Supplier: Thermo Scientific Chemicals A1268014
Description
4-Amino-1-butanol is a useful intermediate in a variety of organic synthesis. It is mainly used in personal-care products, preparation of water soluble cationic flocculants and ion exchange resins as well as in the preparation of beta-lactam antibiotics. It is used for the production of efficient anionic emulsifiers, nonionic polyethylene emulsions, water treatment, metal treatment and absorption of carbon dioxide gas. It is used as a pigment dispersion aid and curing agent in selected textile -resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Amino-1-butanol is a useful intermediate in a variety of organic synthesis. It is mainly used in personal-care products, preparation of water soluble cationic flocculants and ion exchange resins as well as in the preparation of beta-lactam antibiotics. It is used for the production of efficient anionic emulsifiers, nonionic polyethylene emulsions, water treatment, metal treatment and absorption of carbon dioxide gas. It is used as a pigment dispersion aid and curing agent in selected textile -resins.
Solubility
Miscible with water.
Notes
Air Sensitive and hygroscopic. Incompatible with strong oxidizing agents and acids.
Specifications
| 4-Amino-1-butanol | |
| 16°C to 18°C | |
| 206°C | |
| Amine-like | |
| 1.461 | |
| MFCD00008230 | |
| UN2735 | |
| Air sensitive; Hygroscopic | |
| Miscible with water. | |
| C(CCO)CN | |
| 89.138 | |
| 89.14 |
| 13325-10-5 | |
| 0.967 | |
| 107°C (224°F) | |
| C4H11NO | |
| H2N(CH2)4OH | |
| 25 g | |
| 1731411 | |
| 4-amino-1-butanol, 4-aminobutanol, 1-butanol, 4-amino, 4-amino-butan-1-ol, 4-hdyroxybutylamine, 4-hydroxybutylamine, 4-hydroxy-n-butylamine, butanolamine, 4-amino butanol, 4-amino-butanol | |
| BLFRQYKZFKYQLO-UHFFFAOYSA-N | |
| 4-aminobutan-1-ol | |
| 25868 | |
| 98% |
Chemical Identifiers
| 13325-10-5 | |
| 89.138 | |
| BLFRQYKZFKYQLO-UHFFFAOYSA-N | |
| 25868 | |
| C(CCO)CN |
| C4H11NO | |
| MFCD00008230 | |
| 4-amino-1-butanol, 4-aminobutanol, 1-butanol, 4-amino, 4-amino-butan-1-ol, 4-hdyroxybutylamine, 4-hydroxybutylamine, 4-hydroxy-n-butylamine, butanolamine, 4-amino butanol, 4-amino-butanol | |
| 4-aminobutan-1-ol |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 236-364-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only