missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-butanol, 98%

CAS: 13325-10-5 | C4H11NO | 89.138 g/mol

Supplier:  Thermo Scientific Chemicals A1268003

 View more versions of this product

Catalog No. AAA1268003


Only null left
Explore available promotions

Description

Description

4-Amino-1-butanol is a useful intermediate in a variety of organic synthesis. It is mainly used in personal-care products, preparation of water soluble cationic flocculants and ion exchange resins as well as in the preparation of beta-lactam antibiotics. It is used for the production of efficient anionic emulsifiers, nonionic polyethylene emulsions, water treatment, metal treatment and absorption of carbon dioxide gas. It is used as a pigment dispersion aid and curing agent in selected textile -resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-1-butanol is a useful intermediate in a variety of organic synthesis. It is mainly used in personal-care products, preparation of water soluble cationic flocculants and ion exchange resins as well as in the preparation of beta-lactam antibiotics. It is used for the production of efficient anionic emulsifiers, nonionic polyethylene emulsions, water treatment, metal treatment and absorption of carbon dioxide gas. It is used as a pigment dispersion aid and curing agent in selected textile -resins.

Solubility
Miscible with water.

Notes
Air Sensitive and hygroscopic. Incompatible with strong oxidizing agents and acids.
Specifications

Specifications

4-Amino-1-butanol
16°C to 18°C
206°C
Amine-like
1.461
MFCD00008230
UN2735
Air sensitive; Hygroscopic
Miscible with water.
C(CCO)CN
89.138
89.14
13325-10-5
0.967
107°C (224°F)
C4H11NO
H2N(CH2)4OH
1 g
1731411
4-amino-1-butanol, 4-aminobutanol, 1-butanol, 4-amino, 4-amino-butan-1-ol, 4-hdyroxybutylamine, 4-hydroxybutylamine, 4-hydroxy-n-butylamine, butanolamine, 4-amino butanol, 4-amino-butanol
BLFRQYKZFKYQLO-UHFFFAOYSA-N
4-aminobutan-1-ol
25868
98%

Chemical Identifiers

13325-10-5
89.138
BLFRQYKZFKYQLO-UHFFFAOYSA-N
25868
C(CCO)CN
C4H11NO
MFCD00008230
4-amino-1-butanol, 4-aminobutanol, 1-butanol, 4-amino, 4-amino-butan-1-ol, 4-hdyroxybutylamine, 4-hydroxybutylamine, 4-hydroxy-n-butylamine, butanolamine, 4-amino butanol, 4-amino-butanol
4-aminobutan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 236-364-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only